Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORC2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026920.1 | 2 | 1086 | |||
GenBank mRNA | AK093120.1 | - | 2 | 985 | ||
AK098685.1 | - | 1 | 467 | |||
BC042980.1 | - | 3 | 1225 | |||
BC065553.1 | - | 2 | 979 | |||
BC117160.1 | - | 1 | 721 | |||
BC117186.1 | - | 1 | 721 |
Gene Symbol: MORC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BM683680.1 | - | 1 | 493 |