Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX11L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024004.1 | 3 | 584 | |||
GenBank mRNA | AK057285.1 | - | 2 | 590 | ||
AK093685.1 | - | 2 | 1534 | |||
AK123946.1 | - | 3 | 584 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AI908255.1 | - | 2 | 385 | Chr. 2 - 113600535 - 113601358 [-] on Build GRCh38 |