Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX11L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024004.1 | 3 | 939 | |||
GenBank mRNA | AK057285.1 | - | 2 | 942 | ||
AK093685.1 | - | 2 | 1886 | |||
AK123946.1 | - | 3 | 936 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK125998.1 | - | 1 | 2233 | Chr. 9 - 11054 - 14525 [+] on Build GRCh38 |