Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX11L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024004.1 | 3 | 995 | |||
NR_024005.2 | 3 | 498 | ||||
GenBank mRNA | AK057285.1 | - | 2 | 998 | ||
AK093685.1 | - | 2 | 1942 | |||
AK123946.1 | - | 3 | 992 | |||
AM992870.1 | - | 3 | 498 |
Gene Symbol: DDX11L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046018.2 | 3 | 491 | |||
GenBank mRNA | AM992871.1 | - | 3 | 491 |
Gene Symbol: DDX11L10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045117.1 | 3 | 491 | |||
GenBank mRNA | AM992876.1 | - | 3 | 491 |
Gene Symbol: DDX11L11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AM992877.1 | - | 3 | 491 | ||
DA948568.1 | - | 3 | 511 |
Gene Symbol: DDX11L12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AM992878.1 | - | 3 | 434 |
Gene Symbol: DDX11L16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110561.1 | 3 | 491 | |||
GenBank mRNA | AM992882.1 | - | 3 | 491 |
Gene Symbol: DDX11L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AM992872.1 | - | 3 | 491 |
Gene Symbol: DDX11L4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AM992873.1 | - | 2 | 137 |
Gene Symbol: DDX11L5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_051986.1 | 3 | 491 | |||
GenBank mRNA | AM992874.1 | - | 3 | 491 |
Gene Symbol: DDX11L9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034090.1 | 3 | 491 | |||
GenBank mRNA | AM992875.1 | - | 3 | 491 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK125998.1 | - | 1 | 2289 | Chr. 9 - 11054 - 14525 [+] on Build GRCh38 |