Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00950 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024006.1 | 1 | 809 | |||
GenBank mRNA | AF370382.1 | - | 1 | 788 | ||
AL832569.1 | - | 1 | 2341 |
Gene Symbol: TMEM8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011517916.2 | XP_011516218.1 | 9 | 8646 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA231911.1 | - | 1 | 84 | Chr. 9 - 35860999 - 35861577 [+] on Build GRCh38 |