Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00950 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024006.1 | 1 | 112 | |||
GenBank mRNA | AF370382.1 | - | 1 | 91 | ||
AL832569.1 | - | 1 | 1648 |
Gene Symbol: TMEM8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011517916.2 | XP_011516218.1 | 9 | 7949 |