Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027263.1 | 1 | 922 | |||
GenBank mRNA | AF147421.1 | - | 2 | 480 | ||
BC007251.1 | - | 1 | 785 |