Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028041.1 | 6, 7 | 1666 | |||
GenBank mRNA | AJ236922.1 | - | 6, 7 | 1370 | ||
AK123053.1 | - | 7, 8 | 1072 | |||
BC143344.1 | - | 6, 7 | 1381 |