Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP27X-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026742.1 | 2 | 1449 | |||
GenBank mRNA | AK056746.1 | - | 2 | 1335 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000437322.2 | - | 2 | 1334 | Chr. X - 49876724 - 49879241 [-] on Build GRCh38 |