Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP27X-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026742.1 | 2 | 2238 | |||
GenBank mRNA | BI086725.1 | - | 1 | 633 | ||
BX106362.1 | - | 1 | 438 |