Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024247.1 | 4 | 326 | |||
XM_011528385.2 | XP_011526687.1 | 5 | 448 | |||
GenBank mRNA | AK309092.1 | - | 3 | 145 | ||
AK310721.1 | - | 5 | 360 | |||
BC130443.1 | - | 4 | 286 | 40146474 | ||
BC144147.1 | - | 4 | 286 | |||
U20896.1 | - | 2 | 1255 |