Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TINCR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027064.2 | 3 | 2818 | |||
GenBank mRNA | BC036545.1 | - | 3 | 2783 | ||
BC047562.1 | - | 1 | 301 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG354568.1 | - | 1 | 213 | Chr. 19 - 5558175 - 5559294 [-] on Build GRCh38 |