Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SATB2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026830.1 | 2 | 3273 | |||
GenBank mRNA | AK123639.1 | - | 2 | 3286 | ||
AK311461.1 | - | 11 | ||||
BC035902.1 | - | 1 | 2636 |