Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SATB2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026830.1 | 2 | 1105 | |||
GenBank mRNA | AK056625.1 | - | 1 | 652 | ||
AK123639.1 | - | 2 | 1118 | |||
AK290476.1 | - | 1 | 552 | |||
AK291368.1 | - | 2 | 400 | |||
BC035902.1 | - | 1 | 468 | |||
BC101782.1 | - | 1 | 31 | |||
BC113374.1 | - | 1 | 31 |