Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028139.1 | 2 | 2043 | |||
NR_028140.1 | 3 | 451 | ||||
GenBank mRNA | AK057536.1 | - | 2 | 980 | ||
BC028144.1 | - | 3 | 447 | 5214282 | ||
BC132704.1 | - | 3 | 394 | |||
BC132706.1 | - | 3 | 394 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI909450.1 | - | 3 | 447 | Chr. 15 - 100345200 - 100350229 [-] on Build GRCh38 |