Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAVCR1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003603.1 | 1 | 1111 |
Gene Symbol: HAVCR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF043724.1 | - | 9 | 1200 |