Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HPGD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000860.5 | NP_000851.2 | 1 | 235 | ||
NM_001145816.2 | NP_001139288.1 | 1 | 235 | |||
NM_001256301.1 | NP_001243230.1 | 1 | 230 | |||
NM_001256305.1 | NP_001243234.1 | 1 | 235 | |||
NM_001256306.1 | NP_001243235.1 | 1 | 235 | |||
NM_001256307.1 | NP_001243236.1 | 1 | 235 | |||
XM_011531907.1 | XP_011530209.1 | 1 | 230 | |||
GenBank mRNA | AK296642.1 | - | 1 | 230 | ||
DC420578.1 | - | 1 | 235 |