Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSCR9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026719.2 | 3 | 341 | |||
GenBank mRNA | AB066100.1 | - | 1 | 108 | ||
AB212286.1 | - | 3 | 341 | |||
AB212287.1 | - | 4 | 399 | |||
AB212288.1 | - | 5 | 488 | |||
AB212290.1 | - | 2 | 299 | |||
AB212291.1 | - | 4 | 430 | |||
BC066653.1 | - | 3 | 341 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ223675.1 | - | 340 | Chr. - - [-] on Build |