Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DSCR9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026719.2 | 3 | 160 | |||
GenBank mRNA | AB212286.1 | - | 3 | 160 | ||
AB212287.1 | - | 4 | 218 | |||
AB212288.1 | - | 5 | 307 | |||
AB212290.1 | - | 2 | 118 | |||
AB212291.1 | - | 4 | 249 | |||
BC066653.1 | - | 3 | 160 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ223675.1 | - | 159 | Chr. - - [-] on Build |