Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF876P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027481.1 | 2 | 1174 | |||
GenBank mRNA | BC028359.1 | - | 2 | 1169 | ||
BC046475.1 | - | 2 | 1144 |