Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293626.1 | NP_001280555.1 | 11 | 1195 | ||
XM_011516692.2 | XP_011514994.1 | 4 | 410 | |||
GenBank mRNA | AK000645.1 | - | 11 | 1195 | ||
BC111973.1 | - | 11 | 1202 | |||
BC111975.1 | - | 11 | 1202 |