Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MGAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293626.1 | NP_001280555.1 | 6, 7 | 632 | ||
GenBank mRNA | AK000645.1 | - | 6, 7 | 632 | ||
BC111973.1 | - | 6, 7 | 639 | |||
BC111975.1 | - | 6, 7 | 639 |