Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF300P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026867.1 | 5 | 601 | |||
GenBank mRNA | AK091436.1 | - | 5 | 567 | ||
AK096536.1 | - | 5 | 1752 | |||
AK097462.1 | - | 4 | 459 | |||
BC022559.1 | - | 5 | 600 | |||
BC117490.1 | - | 5 | 563 | |||
BC126474.1 | - | 5 | 563 |