Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF300P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026867.1 | 4, 5 | 558 | |||
GenBank mRNA | AK097462.1 | - | 3, 4 | 416 | ||
BC022559.1 | - | 4, 5 | 557 | |||
BC117490.1 | - | 4, 5 | 520 | |||
BC126474.1 | - | 4, 5 | 520 |