Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF300P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026867.1 | 5 | 785 | |||
GenBank mRNA | AK091436.1 | - | 5 | 751 | ||
AK096536.1 | - | 5 | 1936 | |||
AK097462.1 | - | 4 | 643 | |||
BC022559.1 | - | 5 | 784 | |||
BC117490.1 | - | 5 | 747 | |||
BC126474.1 | - | 5 | 747 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CD359466.1 | - | 5 | 704 | Chr. 5 - 150931945 - 150946533 [-] on Build GRCh38 |