Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKRN7P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026640.1 | 1 | 927 |
Gene Symbol: ZNF334 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017027944.1 | XP_016883433.1 | 10 | 5062 |