Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST7-OT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
RefSeq | NR_002332.2 | 6 | 464 | |||
GenBank mRNA | AF400044.1 | - | LIT1900 ST7OT3 |
6 | 464 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU854468.1 | - | 270 | Chr. - - [-] on Build |