Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL23AP87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029406.1 | 3 | 872 | |||
GenBank mRNA | AK125669.1 | - | 3 | 872 |
Gene Symbol: LOC728323 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130699.1 | 8 | 1353 | |||
NR_130700.1 | 6 | 1125 | ||||
NR_130701.1 | 8 | 1278 | ||||
NR_130702.1 | 7 | 1229 | ||||
GenBank mRNA | AK125248.1 | - | 7 | 1166 | ||
AK309231.1 | - | 6 | 1074 |