Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL23AP87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029406.1 | 2 | 380 | |||
GenBank mRNA | AK125669.1 | - | 2 | 380 |
Gene Symbol: LOC101929692 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011536309.1 | XP_011534611.1 | 5 | 1441 |
Gene Symbol: LOC105378947 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011542538.1 | XP_011540840.1 | 5 | 2179 |
Gene Symbol: LOC105379561 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017030172.1 | XP_016885661.1 | 4 | 1156 | ||
XM_017030173.1 | XP_016885662.1 | 3 | 1030 | |||
XM_017030174.1 | XP_016885663.1 | 5 | 752 |