Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL23AP87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029406.1 | 3 | 1223 | |||
GenBank mRNA | AK125669.1 | - | 3 | 1221 |
Gene Symbol: LOC728323 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK125248.1 | - | 7 | 1516 |