Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027244.1 | 1 | 85 | |||
GenBank mRNA | BP341570.1 | - | 1 | 71 | ||
DB205082.1 | - | 1 | 85 |
Gene Symbol: LINC01123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046110.1 | 1 | 74 | |||
NR_046111.1 | 1 | 74 | ||||
NR_046112.1 | 1 | 74 | ||||
GenBank mRNA | BP247062.1 | - | 1 | 74 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AW675296.1 | - | 1 | 68 | Chr. 2 - 109986987 - 109994593 [+] on Build GRCh38 |