Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00493 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015432.1 | 2 | 378 | |||
GenBank mRNA | AF147354.1 | - | 2 | 356 | ||
AK024594.1 | - | 2 | 378 | |||
BC015983.1 | - | 2 | 373 | |||
BC039422.1 | - | 2 | 357 | |||
BC092494.1 | - | 2 | 379 | |||
BC127753.1 | - | 2 | 357 | |||
BC127754.1 | - | 2 | 357 | |||
BC141963.1 | - | 2 | 357 | |||
BC141964.1 | - | 2 | 357 | |||
BC150559.1 | - | 2 | 360 | |||
BC150560.1 | - | 2 | 360 |