Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR99AHG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027790.3 | 8 | 1402 | |||
NR_027791.3 | 6 | 1205 | ||||
NR_111004.2 | 6 | 1271 | ||||
NR_111005.2 | 7 | 1332 | ||||
NR_136542.1 | 8 | 1466 | ||||
NR_136543.1 | 5 | 1144 | ||||
NR_136544.1 | 4 | 1357 | ||||
NR_136545.1 | 5 | 1472 | ||||
NR_136546.1 | 3 | 1296 | ||||
NR_136547.1 | 3 | 907 | ||||
NR_136548.1 | 4 | 1161 | ||||
NR_136549.1 | 4 | 1088 | ||||
NR_136550.1 | 4 | 1096 | ||||
NR_136551.1 | 6 | 1240 | ||||
NR_136552.1 | 7 | 1355 | ||||
GenBank mRNA | AK095614.1 | - | 8 | 1402 |