Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR99AHG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027790.3 | 8 | 1137 | |||
NR_027791.3 | 6 | 940 | ||||
NR_111004.2 | 6 | 1006 | ||||
NR_111005.2 | 7 | 1067 | ||||
NR_136542.1 | 8 | 1201 | ||||
NR_136543.1 | 5 | 879 | ||||
NR_136544.1 | 4 | 1092 | ||||
NR_136545.1 | 5 | 1207 | ||||
NR_136546.1 | 3 | 1031 | ||||
NR_136547.1 | 3 | 642 | ||||
NR_136548.1 | 4 | 896 | ||||
NR_136549.1 | 4 | 823 | ||||
NR_136550.1 | 4 | 831 | ||||
NR_136551.1 | 6 | 975 | ||||
NR_136552.1 | 7 | 1090 | ||||
GenBank mRNA | AK095614.1 | - | 8 | 1137 |