Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR99AHG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027790.3 | 8 | 1001 | |||
NR_027791.3 | 6 | 804 | ||||
NR_111004.2 | 6 | 870 | ||||
NR_111005.2 | 7 | 931 | ||||
NR_136542.1 | 8 | 1065 | ||||
NR_136543.1 | 5 | 743 | ||||
NR_136544.1 | 4 | 956 | ||||
NR_136545.1 | 5 | 1071 | ||||
NR_136546.1 | 3 | 895 | ||||
NR_136547.1 | 3 | 506 | ||||
NR_136548.1 | 4 | 760 | ||||
NR_136549.1 | 4 | 687 | ||||
NR_136550.1 | 4 | 695 | ||||
NR_136551.1 | 6 | 839 | ||||
NR_136552.1 | 7 | 954 | ||||
GenBank mRNA | AK095614.1 | - | 8 | 1001 |