Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR99AHG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027790.3 | 8 | 2677 | |||
NR_027791.3 | 6 | 2480 | ||||
NR_111004.2 | 6 | 2546 | ||||
NR_111005.2 | 7 | 2607 | ||||
NR_136542.1 | 8 | 2741 | ||||
NR_136543.1 | 5 | 2419 | ||||
NR_136544.1 | 4 | 2632 | ||||
NR_136545.1 | 5 | 2747 | ||||
NR_136546.1 | 3 | 2571 | ||||
NR_136547.1 | 3 | 2182 | ||||
NR_136548.1 | 4 | 2436 | ||||
NR_136549.1 | 4 | 2363 | ||||
NR_136550.1 | 4 | 2371 | ||||
NR_136551.1 | 6 | 2515 | ||||
NR_136552.1 | 7 | 2630 | ||||
GenBank mRNA | AK095614.1 | - | 8 | 2677 |