Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR99AHG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027790.3 | 8 | 1567 | |||
NR_027791.3 | 6 | 1370 | ||||
NR_111004.2 | 6 | 1436 | ||||
NR_111005.2 | 7 | 1497 | ||||
NR_136542.1 | 8 | 1631 | ||||
NR_136543.1 | 5 | 1309 | ||||
NR_136544.1 | 4 | 1522 | ||||
NR_136545.1 | 5 | 1637 | ||||
NR_136546.1 | 3 | 1461 | ||||
NR_136547.1 | 3 | 1072 | ||||
NR_136548.1 | 4 | 1326 | ||||
NR_136549.1 | 4 | 1253 | ||||
NR_136550.1 | 4 | 1261 | ||||
NR_136551.1 | 6 | 1405 | ||||
NR_136552.1 | 7 | 1520 | ||||
GenBank mRNA | AK095614.1 | - | 8 | 1567 |