Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR99AHG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027790.3 | 8 | 3180 | |||
NR_027791.3 | 6 | 2983 | ||||
NR_111004.2 | 6 | 3049 | ||||
NR_111005.2 | 7 | 3110 | ||||
NR_136542.1 | 8 | 3244 | ||||
NR_136543.1 | 5 | 2922 | ||||
NR_136544.1 | 4 | 3135 | ||||
NR_136545.1 | 5 | 3250 | ||||
NR_136546.1 | 3 | 3074 | ||||
NR_136547.1 | 3 | 2685 | ||||
NR_136548.1 | 4 | 2939 | ||||
NR_136549.1 | 4 | 2866 | ||||
NR_136550.1 | 4 | 2874 | ||||
NR_136551.1 | 6 | 3018 | ||||
NR_136552.1 | 7 | 3133 | ||||
GenBank mRNA | AK095614.1 | - | 8 | 3180 |