Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: A1BG-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015380.2 | 4 | 1487 | |||
GenBank mRNA | AK027222.1 | - | 4 | 1487 | ||
BC006144.1 | - | 1 | 319 | |||
BC040926.1 | - | 3 | 1075 |
Gene Symbol: ZNF497 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CA438336.1 | - | 1 | 200 |