Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SERHL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027786.1 | 1 | 93 | |||
GenBank mRNA | AK094872.1 | - | 1 | 93 | ||
AL365513.1 | - | 1 | 82 | |||
BC093888.1 | - | 1 | 47 | 7939733 | ||
DA273263.1 | - | 1 | 93 |