Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNORA16A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003035.1 | 1 | 62 | |||
GenBank mRNA | AJ609433.1 | - | 1 | 62 |
Gene Symbol: SNHG12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK092096.1 | - | 1 | 884 | ||
BC034611.1 | - | 2 | 313 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV697366.1 | - | 1 | 49 | Chr. 1 - 28579532 - 28581040 [-] on Build GRCh38 |