Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGALS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003225.2 | 1 | 980 | |||
GenBank mRNA | AB209391.1 | - | 2 | 4067 | ||
AF266280.1 | - | 1 | 307 | |||
BC128253.1 | - | 1 | 759 | |||
BC128403.1 | - | 1 | 759 | |||
BX161455.1 | - | 1 | 775 | |||
BX641090.1 | - | 1 | 981 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX388444.2 | - | 1 | 780 | Chr. 14 - 55137183 - 55138181 [+] on Build GRCh38 |