Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGALS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003225.2 | 1 | 374 | |||
GenBank mRNA | AB209391.1 | - | 2 | 3461 | ||
BC128253.1 | - | 1 | 153 | |||
BC128403.1 | - | 1 | 153 | |||
BX161455.1 | - | 1 | 169 | |||
BX641090.1 | - | 1 | 375 |