Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRCA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027676.1 | 6, 7 | 577 | |||
GenBank mRNA | AF005068.1 | - | 6, 7 | 542 | ||
BC085615.1 | - | 6, 7 | 540 |