Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDRT15P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003261.1 | 1 | 32 | |||
GenBank mRNA | AA453375.1 | - | 1 | 34 | ||
BX118788.1 | - | 1 | 33 |