Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARGP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029388.2 | 5, 6 | 635 | |||
GenBank mRNA | AK311609.1 | - | 5, 6 | 607 |
Gene Symbol: PARG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136754.1 | 7, 8 | 2052 | |||
NR_136755.1 | 8, 9 | 2119 | ||||
NR_136756.1 | 8, 9 | 2119 |