Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM23B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290117.1 | NP_001277046.1 | 7 | 777 | ||
NM_001290118.1 | NP_001277047.1 | 5 | 633 | |||
NR_110767.1 | 8 | 901 | ||||
NR_135475.1 | 7 | 777 | ||||
GenBank mRNA | AK293228.1 | - | 5 | 585 | ||
AK295227.1 | - | 7 | 1087 | |||
AK295370.1 | - | 7 | 729 | |||
AK316490.1 | - | 7 | 729 | |||
AK316549.1 | - | 7 | 778 | |||
BC035067.1 | - | 2 | 655 |
Gene Symbol: PARGP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029388.2 | 12 | 1357 | |||
GenBank mRNA | AF130105.1 | - | 1 | 1687 | ||
AK293897.1 | - | 8 | 885 | |||
AK311609.1 | - | 12 | 1329 |