Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARGP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029388.2 | 4 | 500 | |||
GenBank mRNA | AK311609.1 | - | 4 | 472 | ||
DA800072.1 | - | 4 | 500 |