Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GHRLOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_004431.3 | 5 | 1051 | |||
NR_024144.2 | 6 | 879 | ||||
NR_024145.2 | 6 | 1223 | ||||
NR_073566.1 | 7 | 1234 | ||||
NR_073567.1 | 6 | 1222 | ||||
NR_073568.1 | 6 | 1077 | ||||
GenBank mRNA | BX118778.1 | - | 1 | 212 | ||
EF549568.1 | - | 4 | 994 | |||
EF549570.1 | - | 4 | 739 | |||
EU072081.1 | - | 3 | 926 | |||
EU072082.1 | - | 2 | 768 | |||
EU789531.1 | - | 3 | 545 | |||
EU789533.1 | - | 1 | 93 |