Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEIS3P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002211.1 | 1 | 1572 | |||
GenBank mRNA | AK054814.1 | - | 1 | 1571 | ||
U68385.1 | - | 1 | 1357 |